COM448620748 Combi-Blocks, Inc.

3-(2-Bromophenyl)-5-(3-nitrophenyl)-1,2,4-oxadiazole

  • MDL number MFCD09878377
  • Cas Number : 1000339-27-4
  • Molecular Weight : 346.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-6288

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-6288
Availability details for
Quantity
  • Availability
  • Add to Cart