COM448620792 Combi-Blocks, Inc.

6-Bromo-2-cyclohexylquinazolin-4(3H)-one

  • MDL number MFCD09878379
  • Cas Number : 1000339-29-6
  • Molecular Weight : 307.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 98

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?HC-6360
Availability details for
Quantity
  • Availability
  • Add to Cart