COM448620840 Combi-Blocks, Inc.

5-Cyclohexyl-3-p-tolyl-1,2,4-oxadiazole

  • MDL number MFCD02214602
  • Cas Number : 330828-28-9
  • Molecular Weight : 242.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-6423

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-6423
Availability details for
Quantity
  • Availability
  • Add to Cart