COM448620906 Combi-Blocks, Inc.

5-Ethyl-3-(4-nitrophenyl)-1,2,4-oxadiazole

  • MDL number MFCD09972166
  • Cas Number : 10185-65-6
  • Molecular Weight : 219.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-6505

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-6505
Availability details for
Quantity
  • Availability
  • Add to Cart