COM448621069 Combi-Blocks, Inc.

2-Nitroacetamide

  • MDL number MFCD01215007
  • Cas Number : 14011-21-3
  • Molecular Weight : 104.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-6800

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-6800
Availability details for
Quantity
  • Availability
  • Add to Cart