COM448621104 Combi-Blocks, Inc.

3-(3-Chlorophenyl)-1H-pyrazole

  • MDL number MFCD09860599
  • Cas Number : 59843-69-5
  • Molecular Weight : 178.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-6892

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-6892
Availability details for
Quantity
  • Availability
  • Add to Cart