COM448621207 Combi-Blocks, Inc.

4-Iodo-1-methyl-1H-imidazole-5-carboxaldehyde

  • MDL number MFCD01632219
  • Cas Number : 189014-13-9
  • Molecular Weight : 236.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HI-1028

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HI-1028
Availability details for
Quantity
  • Availability
  • Add to Cart