COM448621249 Combi-Blocks, Inc.

4-Bromo-2-methylimidazole

  • MDL number MFCD03411959
  • Cas Number : 16265-11-5
  • Molecular Weight : 161.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HI-1111

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HI-1111
Availability details for
Quantity
  • Availability
  • Add to Cart