COM448621277 Combi-Blocks, Inc.

2-Bromo-1-methyl-1H-imidazole, HCl

  • MDL number MFCD08235099
  • Cas Number : 913836-21-2
  • Molecular Weight : 197.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HI-1148

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HI-1148
Availability details for
Quantity
  • Availability
  • Add to Cart