COM448621369 Combi-Blocks, Inc.

1-(4-fluorophenyl)pyrazole

  • MDL number MFCD12192639
  • Cas Number : 81329-32-0
  • Molecular Weight : 162.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HI-1283

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?HI-1283
Availability details for
Quantity
  • Availability
  • Add to Cart