COM448621429 Combi-Blocks, Inc.

5-Bromo-1-(4-methoxybenzyl)-1H-benzo[d]imidazole

  • MDL number MFCD09972169
  • Cas Number : 1020253-11-5
  • Molecular Weight : 317.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HI-1371

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HI-1371
Availability details for
Quantity
  • Availability
  • Add to Cart