COM448621446 Combi-Blocks, Inc.

5-Amino-4-cyano-1-(2,4-dimethylphenyl)-3-methylpyrazole

  • MDL number MFCD10699684
  • Cas Number : 1072944-82-1
  • Molecular Weight : 226.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HI-1403

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HI-1403
Availability details for
Quantity
  • Availability
  • Add to Cart