COM448621509 Combi-Blocks, Inc.

4-Bromo-1-isopentylpyrazole

  • MDL number MFCD12026053
  • Cas Number : 847818-48-8
  • Molecular Weight : 217.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HI-1507

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HI-1507
Availability details for
Quantity
  • Availability
  • Add to Cart