COM448621893 Combi-Blocks, Inc.

Ethyl 2-(2-bromophenyl)-5-methylpyrazole-3-carboxylate

  • MDL number MFCD27935907
  • Cas Number : 1448684-44-3
  • Molecular Weight : 309.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2087

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2087
Availability details for
Quantity
  • Availability
  • Add to Cart