COM448621899 Combi-Blocks, Inc.

1-(Oxan-2-yl)pyrazole-3-carbaldehyde

  • MDL number MFCD27981310
  • Cas Number : 1809564-49-5
  • Molecular Weight : 180.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2111

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2111
Availability details for
Quantity
  • Availability
  • Add to Cart