COM448621902 Combi-Blocks, Inc.

(1-Benzylindazol-3-yl)methanol

  • MDL number MFCD01631172
  • Cas Number : 131427-21-9
  • Molecular Weight : 238.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2115

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2115
Availability details for
Quantity
  • Availability
  • Add to Cart