COM448621907 Combi-Blocks, Inc.

N-(N-BOC-Piperidino)pyrazole-4-carboxaldehyde

  • MDL number MFCD19687734
  • Cas Number : 1076224-00-4
  • Molecular Weight : 279.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2120

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2120
Availability details for
Quantity
  • Availability
  • Add to Cart