COM448621914 Combi-Blocks, Inc.

4-Methoxy-1,3-benzothiazole

  • MDL number MFCD16877170
  • Cas Number : 3048-46-2
  • Molecular Weight : 165.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2131

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2131
Availability details for
Quantity
  • Availability
  • Add to Cart