COM448621915 Combi-Blocks, Inc.

N-(2-Hydroxyethyl)-1H-pyrrole-2-carboxamide

  • MDL number MFCD12179285
  • Cas Number : 1153189-07-1
  • Molecular Weight : 154.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2132

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2132
Availability details for
Quantity
  • Availability
  • Add to Cart