COM448621918 Combi-Blocks, Inc.

2-(Benzyloxy)-1,3-thiazole

  • MDL number MFCD28096635
  • Cas Number : 1304771-28-5
  • Molecular Weight : 191.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2135

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2135
Availability details for
Quantity
  • Availability
  • Add to Cart