COM448621919 Combi-Blocks, Inc.

1-Propyl-3-(trifluoromethyl)pyrazole-4-carboxylic acid

  • MDL number MFCD28024841
  • Cas Number : 1103427-26-4
  • Molecular Weight : 222.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2137

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2137
Availability details for
Quantity
  • Availability
  • Add to Cart