COM448621922 Combi-Blocks, Inc.

4-(Pyrrolidin-1-ylmethyl)benzonitrile

  • MDL number MFCD07325398
  • Cas Number : 78064-96-7
  • Molecular Weight : 186.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2140

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2140
Availability details for
Quantity
  • Availability
  • Add to Cart