COM448621925 Combi-Blocks, Inc.

4-(Pyrazol-1-ylmethyl)benzonitrile

  • MDL number MFCD07687291
  • Cas Number : 179057-34-2
  • Molecular Weight : 183.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2143

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2143
Availability details for
Quantity
  • Availability
  • Add to Cart