COM448621926 Combi-Blocks, Inc.

Methyl 4-cyanothiophene-2-carboxylate

  • MDL number MFCD05664843
  • Cas Number : 67808-33-7
  • Molecular Weight : 167.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2149

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2149
Availability details for
Quantity
  • Availability
  • Add to Cart