COM448621934 Combi-Blocks, Inc.

2-(2-Bromophenyl)-5-methylpyrazole-3-carboxylic acid

  • MDL number MFCD26611809
  • Cas Number : 1448684-51-2
  • Molecular Weight : 281.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2162

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2162
Availability details for
Quantity
  • Availability
  • Add to Cart