COM448621937 Combi-Blocks, Inc.

1-[3-(Trifluoromethyl)pyrazol-1-yl]propan-2-ol

  • MDL number MFCD16065387
  • Cas Number : 742096-93-1
  • Molecular Weight : 194.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Refrigerated

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2165
Availability details for
Quantity
  • Availability
  • Add to Cart