COM448621938 Combi-Blocks, Inc.

tert-Butyl 2-(4-bromophenyl)imidazole-1-carboxylate

  • MDL number MFCD26390250
  • Cas Number : 176961-27-6
  • Molecular Weight : 323.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2167

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2167
Availability details for
Quantity
  • Availability
  • Add to Cart