COM448621940 Combi-Blocks, Inc.

1-[5-(Trifluoromethyl)pyrazol-1-yl]propan-2-ol

  • MDL number MFCD28127186
  • Cas Number : 1820707-72-9
  • Molecular Weight : 194.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2169

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2169
Availability details for
Quantity
  • Availability
  • Add to Cart