COM448621943 Combi-Blocks, Inc.

2-Bromo-4-(dimethoxymethyl)-1,3-thiazole

  • MDL number MFCD28143934
  • Cas Number : 1313399-79-9
  • Molecular Weight : 238.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2174

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2174
Availability details for
Quantity
  • Availability
  • Add to Cart