COM448621944 Combi-Blocks, Inc.

Methyl 6-methyl-4-oxo-6H,7H-pyrazolo[3,2-c][1,4]oxazine-2-carboxylate

  • MDL number MFCD28143935
  • Cas Number : 1820686-06-3
  • Molecular Weight : 210.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2177

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2177
Availability details for
Quantity
  • Availability
  • Add to Cart