COM448621945 Combi-Blocks, Inc.

tert-Butyl 4-methylpyrazole-1-carboxylate

  • MDL number MFCD24466467
  • Cas Number : 121669-69-0
  • Molecular Weight : 182.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 97
Storage temp. Refrigerated

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2179
Availability details for
Quantity
  • Availability
  • Add to Cart