COM448621946 Combi-Blocks, Inc.

4,5-Dimethyl-2H-pyrazol-3-amine hydrochloride

  • MDL number MFCD13186039
  • Cas Number : 91159-73-8
  • Molecular Weight : 147.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2180

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2180
Availability details for
Quantity
  • Availability
  • Add to Cart