COM448621949 Combi-Blocks, Inc.

2-[4-(Trifluoromethyl)phenyl]-1,3,4-oxadiazole

  • MDL number MFCD01814630
  • Cas Number : 477886-85-4
  • Molecular Weight : 214.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2183

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2183
Availability details for
Quantity
  • Availability
  • Add to Cart