COM448621954 Combi-Blocks, Inc.

3-(4-Bromophenyl)-4,5-dihydro-1,2-oxazole

  • MDL number MFCD26395060
  • Cas Number : 51067-06-2
  • Molecular Weight : 226.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 98
Storage temp. Refrigerated

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2196
Availability details for
Quantity
  • Availability
  • Add to Cart