COM448621955 Combi-Blocks, Inc.

tert-Butyl 2-phenyl-4H,6H,7H-pyrazolo[4,3-c]pyridine-5-carboxylate

  • MDL number MFCD22384519
  • Cas Number : 1310796-20-3
  • Molecular Weight : 299.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2197

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2197
Availability details for
Quantity
  • Availability
  • Add to Cart