COM448621958 Combi-Blocks, Inc.

4-Bromo-2-(1H-pyrazol-4-yl)aniline

  • MDL number MFCD28397269
  • Cas Number : 1820651-10-2
  • Molecular Weight : 238.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2200

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2200
Availability details for
Quantity
  • Availability
  • Add to Cart