COM448621969 Combi-Blocks, Inc.

2-fluoro-6-(1H-pyrazol-4-yl)aniline

  • MDL number MFCD28397279
  • Cas Number : 1820666-38-3
  • Molecular Weight : 177.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2211

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2211
Availability details for
Quantity
  • Availability
  • Add to Cart