COM448621976 Combi-Blocks, Inc.

3-Benzoylpyrrole

  • MDL number MFCD02179582
  • Cas Number : 7126-41-2
  • Molecular Weight : 171.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HP-1517

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HP-1517
Availability details for
Quantity
  • Availability
  • Add to Cart