COM448622039 Combi-Blocks, Inc.

4,7-Dimethoxyindole

  • MDL number MFCD00178512
  • Cas Number : 23876-39-3
  • Molecular Weight : 177.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?IN-0061

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?IN-0061
Availability details for
Quantity
  • Availability
  • Add to Cart