COM448622309 Combi-Blocks, Inc.

4-Bromo-7-(dibromomethyl)benzo[c][1,2,5]thiadiazole

  • MDL number MFCD22689100
  • Cas Number : 1239277-96-3
  • Molecular Weight : 386.89

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Freezer
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?IN-0431

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?IN-0431
Availability details for
Quantity
  • Availability
  • Add to Cart