COM448622335 Combi-Blocks, Inc.

2-Ethyl-5-nitroindazole

  • MDL number MFCD27981313
  • Cas Number : 5228-50-2
  • Molecular Weight : 191.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?IN-0466

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?IN-0466
Availability details for
Quantity
  • Availability
  • Add to Cart