COM448622337 Combi-Blocks, Inc.

3-Iodo-1H-indazole-5-carbonitrile

  • MDL number MFCD09263216
  • Cas Number : 944898-90-2
  • Molecular Weight : 269.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Refrigerated

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?IN-0469
Availability details for
Quantity
  • Availability
  • Add to Cart