COM448622339 Combi-Blocks, Inc.

tert-Butyl 2-methylindole-1-carboxylate

  • MDL number MFCD16036607
  • Cas Number : 98598-96-0
  • Molecular Weight : 231.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?IN-0472

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?IN-0472
Availability details for
Quantity
  • Availability
  • Add to Cart