COM448622345 Combi-Blocks, Inc.

4-Cyanogramine

  • MDL number MFCD11112015
  • Cas Number : 105907-63-9
  • Molecular Weight : 199.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?IN-0480

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?IN-0480
Availability details for
Quantity
  • Availability
  • Add to Cart