COM448622348 Combi-Blocks, Inc.

6-Bromogramine

  • MDL number MFCD11048384
  • Cas Number : 59609-63-1
  • Molecular Weight : 253.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?IN-0483

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?IN-0483
Availability details for
Quantity
  • Availability
  • Add to Cart