COM448622353 Combi-Blocks, Inc.

3-(Dimethylamino)-7-nitro-1H-indole

  • MDL number MFCD01718873
  • Cas Number : 1654-34-8
  • Molecular Weight : 219.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?IN-0488

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?IN-0488
Availability details for
Quantity
  • Availability
  • Add to Cart