COM448622355 Combi-Blocks, Inc.

1-Methyl-6-nitroindole

  • MDL number MFCD13178347
  • Cas Number : 99459-48-0
  • Molecular Weight : 176.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?IN-0491

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?IN-0491
Availability details for
Quantity
  • Availability
  • Add to Cart