COM448622367 Combi-Blocks, Inc.

Ethyl 3-amino-4-methoxy-1-benzothiophene-2-carboxylate

  • MDL number MFCD13194590
  • Cas Number : 77373-52-5
  • Molecular Weight : 251.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?IN-0508

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?IN-0508
Availability details for
Quantity
  • Availability
  • Add to Cart