COM448622371 Combi-Blocks, Inc.

5-(Trifluoromethoxy)-3a,7a-dihydro-1H-indazol-3-amine

  • MDL number MFCD28369583
  • Cas Number : 1982241-78-0
  • Molecular Weight : 219.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?IN-0513

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?IN-0513
Availability details for
Quantity
  • Availability
  • Add to Cart