COM448622438 Combi-Blocks, Inc.

4-Acetyl-3'-fluoro-4'-methylbiphenyl

  • MDL number MFCD20441875
  • Cas Number : 1345471-18-2
  • Molecular Weight : 228.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-0067

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-0067
Availability details for
Quantity
  • Availability
  • Add to Cart